CAS RN 168316-95-8
IUPAC Name:
13-{[5-(dimethylamino)-6-methyloxan-2-yl]oxy}-9-ethyl-14-methyl-2-[(2,3,4-trimethoxy-5-methylcyclohexyl)oxy]-icosahydro-1H-as-indaceno[3,2-d]oxacyclododecane-7,15-dione
Synonyms:
spinosad
Molecular Weight (g/mol):
734.03
Molecular Formula:
C42H71NO9
InChi Key:
RQOIAWYOVOXMST-UHFFFAOYNA-N
SMILES:
CCC1CCCC(OC2CCC(C(C)O2)N(C)C)C(C)C(=O)C2CC3C4CC(CC4CCC3C2CC(=O)O1)OC1CC(C)C(OC)C(OC)C1OC