accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COONOCH3(E/Z)(E/Z)

CAS RN 16833-17-3

IUPAC Name: butyl 3-(4-{[(4-methoxyphenyl)methylidene]amino}phenyl)prop-2-enoate
Synonyms: butyl 3-(4-{[(4-methoxyphenyl)methylidene]amino}phenyl)prop-2-enoate
Molecular Weight (g/mol): 337.42
Molecular Formula: C21H23NO3
InChi Key: CULYZFOOJBHBNM-UHFFFAOYSA-N
SMILES: CCCCOC(=O)C=CC1=CC=C(C=C1)N=CC1=CC=C(OC)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results