Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butyl 4-[(4-Methoxybenzylidene)amino]cinnamate 98.0+%, TCI America™
SDP

Catalog No. B02551G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B02551G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B02551G Supplier TCI America Supplier No. B02551G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 16833-17-3
Molecular Formula C21H23NO3
Molecular Weight (g/mol) 337.419
MDL Number MFCD00059871
InChI Key CULYZFOOJBHBNM-CIZPVDQASA-N
Synonym 4-(p-Anisalamino)cinnamic Acid Butyl Ester, 4-[(p-Anisylidene)amino]cinnamic Acid Butyl Ester, Butyl 4-(p-Anisalamino)cinnamate, Butyl 4-[(p-Anisylidene)amino]cinnamate, 4-[(4-Methoxybenzylidene)amino]cinnamic Acid Butyl Ester
PubChem CID 5705753
IUPAC Name butyl (E)-3-[4-[(4-methoxyphenyl)methylideneamino]phenyl]prop-2-enoate
SMILES CCCCOC(=O)C=CC1=CC=C(C=C1)N=CC2=CC=C(C=C2)OC

Specifications

Color Yellow
Quantity 1 g
Formula Weight 337.42
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Butyl 4-[(4-Methoxybenzylidene)amino]cinnamate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.