accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOONN

CAS RN 16837-38-0

IUPAC Name: pyridine-3-carbonyl pyridine-3-carboxylate
Synonyms: pyridine-3-carbonyl pyridine-3-carboxylate
Molecular Weight (g/mol): 228.21
Molecular Formula: C12H8N2O3
InChi Key: VPODXHOUBDCEHN-UHFFFAOYSA-N
SMILES: O=C(OC(=O)C1=CC=CN=C1)C1=CC=CN=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results