Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Pyridinecarboxylic Anhydride 97.0+%, TCI America™
SDP

Catalog No. P17685G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P17685G 5 g
P17681G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P17685G Supplier TCI America Supplier No. P17685G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 16837-38-0
Molecular Formula C12H8N2O3
Molecular Weight (g/mol) 228.207
InChI Key VPODXHOUBDCEHN-UHFFFAOYSA-N
Synonym Nicotinic Anhydride
PubChem CID 85607
IUPAC Name pyridine-3-carbonyl pyridine-3-carboxylate
SMILES C1=CC(=CN=C1)C(=O)OC(=O)C2=CN=CC=C2

Specifications

Melting Point 124°C
Color White-Yellow
Boiling Point 200°C
Quantity 5 g
Formula Weight 228.21
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Pyridinecarboxylic Anhydride
EINECSNumber (5)-0740
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.