accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClNH2N

CAS RN 1684-40-8

IUPAC Name: hydrogen 1,2,3,4-tetrahydroacridin-9-amine chloride
Synonyms: hydrogen tacrine chloride
Molecular Weight (g/mol): 234.73
Molecular Formula: C13H15ClN2
InChi Key: ZUFVXZVXEJHHBN-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NC1=C2C=CC=CC2=NC2=C1CCCC2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results