accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16871-60-6

ClClClClClClKKOs

CAS RN 16871-60-6

IUPAC Name: osmium(4+) dipotassium hexachloride
Synonyms: osmium(4+) dipotassium hexachloride
Molecular Weight (g/mol): 481.13
Molecular Formula: Cl6K2Os
InChi Key: VGKQJCSDERXWRV-UHFFFAOYSA-H
SMILES: [Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[K+].[K+].[Os+4]
Quantity 
  • (1)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results