accessibility menu, dialog, popup

UserName

Viewing profile as user:
A(a)

CAS RN 16872-09-6

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 144.25
Molecular Formula: C2H12B10
InChi Key: —
SMILES: —
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)