Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Carborane, 98%, Thermo Scientific Chemicals

Catalog No. AA3513104
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3513104 2 g
AA3513109 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3513104 Supplier Thermo Scientific Chemicals Supplier No. 03513104
Add to Cart

missing translation for 'validMainframeMsg'

o-Carborane is used in a wide range of applications such as heat-resistant polymers and medical applications. In coordination chemistry, it is used as unique bulky ligand scaffolds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16872-09-6
Molecular Formula C2H12B10
Molecular Weight (g/mol) 144.25
MDL Number MFCD00012169
InChI Key WMLCLVOTMCESJH-UHFFFAOYNA-N
Synonym o-carborane, 1,2-dicarbadodecaborane 12, o-carborane 500mg, 11z-1,2,3,4,5,6,7,8,9,10-decaboracyclododec-11-ene
PubChem CID 17750431
IUPAC Name (7R,12R)-1λ⁵,2λ⁵,3λ⁵,4λ⁵,5λ⁵-pentabora-7λ⁴-bora-8λ⁵,10λ⁵-dibora-11λ⁴,12λ⁴-diboraheptadecacyclo[5.5.0.0¹,³.0¹,⁴.0²,¹⁰.0²,¹¹.0²,¹².0³,⁵.0³,¹⁰.0⁴,⁶.0⁴,⁷.0⁵,⁹.0⁵,¹⁰.0⁶,⁸.0⁶,⁹.0⁸,¹¹.0⁸,¹²]dodecane
SMILES [B@@]123[B]45[B]116[B@@]78[B]229[B]343[B]545C11[B]44%10[B]72([B]9354)[C]681%10

Specifications

Melting Point 285°C to 287°C
Color White
Density 0.95 g/mL
Quantity 2 g
UN Number UN1325
Solubility Information Insoluble in water.
Formula Weight 144.25
Percent Purity 98%
Physical Form Powder
Chemical Name or Material o-Carborane
Hazard Category H302
Hazard Statement GHS H Statement
H228-H302
Flammable solid.
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
EINECSNumber 240-897-8
RTECSNumber HS9285000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.