accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OONOO

CAS RN 16881-39-3

IUPAC Name: 1-benzyl 2-tert-butyl pyrrolidine-1,2-dicarboxylate
Synonyms: 1-benzyl 2-tert-butyl pyrrolidine-1,2-dicarboxylate
Molecular Weight (g/mol): 305.37
Molecular Formula: C17H23NO4
InChi Key: OULMZZGGALAOLR-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)C1CCCN1C(=O)OCC1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results