Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Carbobenzoxy-L-proline tert-Butyl Ester 98.0+%, TCI America™
SDP

Catalog No. C17645G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C17645G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C17645G Supplier TCI America Supplier No. C17645G

Chemical Identifiers

CAS 16881-39-3
Molecular Formula C17H23NO4
Molecular Weight (g/mol) 305.374
MDL Number MFCD00051777
InChI Key OULMZZGGALAOLR-AWEZNQCLSA-N
Synonym N-Cbz-L-proline tert-Butyl Ester, Z-Pro-OtBu
PubChem CID 7019799
IUPAC Name 1-O-benzyl 2-O-tert-butyl (2S)-pyrrolidine-1,2-dicarboxylate
SMILES CC(C)(C)OC(=O)C1CCCN1C(=O)OCC2=CC=CC=C2

Specifications

Melting Point 47°C
Color White
Boiling Point 51°C
Quantity 5 g
Formula Weight 305.37
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxy-L-proline tert-Butyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.