accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16881-77-9

IUPAC Name: dimethoxy(methyl)silyl
Synonyms: dimethoxy(methyl)silyl
Molecular Weight (g/mol): 105.19
Molecular Formula: C3H9O2Si
InChi Key: PKTOVQRKCNPVKY-UHFFFAOYSA-N
SMILES: CO[Si](C)OC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results