Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethoxy(methyl)silane 97.0+%, TCI America™
SDP

Catalog No. D210025ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D210025ML 25 mL
D2100100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D210025ML Supplier TCI America Supplier No. D210025ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 16881-77-9
Molecular Formula C3H9O2Si
Molecular Weight (g/mol) 105.188
MDL Number MFCD00048019
InChI Key PKTOVQRKCNPVKY-UHFFFAOYSA-N
Synonym methyldimethoxysilane, dimethoxy methyl silane, silane, dimethoxymethyl, dimethoxy methyl silicon, methyl dimethoxysilane, c3h10o2si, methyl dimethoxy silane, dimethoxy methyl silane #, dimethoxy methyl silyl
PubChem CID 6329250
IUPAC Name dimethoxy(methyl)silicon
SMILES CO[Si](C)OC

Specifications

Color Colorless
Boiling Point 61°C
Quantity 25 mL
UN Number 1992
Formula Weight 106.20
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethoxy(methyl)silane
EINECSNumber (2)-2052&(2)-2053
RTECSNumber VV3644166
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.