accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16903-35-8

IUPAC Name: gold(3+) ammonium tetrachloride
Synonyms: chloroauric acid
Molecular Weight (g/mol): 356.81
Molecular Formula: AuCl4H4N
InChi Key: WPEJSSRSFRWYJB-UHFFFAOYSA-K
SMILES: [NH4+].[Cl-].[Cl-].[Cl-].[Cl-].[Au+3]
Grade 
  • (1)
  • (8)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (8)
  • (2)
  • (2)
  • (1)
  • (8)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)