Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gold Chloride, Trihydrate, ACS Grade, ≥ 49.0 % Au, LabChem™

Catalog No. LC148937 Shop All LabChem Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Vial
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC148937 1 g Vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC148937 Supplier LabChem, Inc. Supplier No. LC148937
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 16903-35-8 , 64-17-5 , 7732-18-5 , 67-63-0 , 67-56-1 , 9000-29-7
Molecular Formula AuCl4H4N
Molecular Weight (g/mol) 356.81
InChI Key WPEJSSRSFRWYJB-UHFFFAOYSA-K
PubChem CID 10925836  
IUPAC Name gold(3+) ammonium tetrachloride
SMILES [NH4+].[Cl-].[Cl-].[Cl-].[Cl-].[Au+3]

Specifications

Melting Point 254°C
Color Golden
Physical Form Lump
Quantity 1 g
Linear Formula HAuCl4·3H2O
Solubility Information Soluble in water
Formula Weight 393.83
Percent Purity ≥49.0% Au
Grade ACS
Identification Passes Test
Packaging Vial
Decomposition Information Hydrogen chloride; Thermal decomposition generates: Corrosive vapors
Chemical Name or Material Gold Chloride, Trihydrate
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Refrigerate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.