accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16911-89-0

IUPAC Name: (phenylsulfanyl)methanecarbothioyl chloride
Synonyms: (phenylsulfanyl)methanecarbothioyl chloride
Molecular Weight (g/mol): 188.69
Molecular Formula: C7H5ClS2
InChi Key: XEXIHCWFFNGQDE-UHFFFAOYSA-N
SMILES: ClC(=S)SC1=CC=CC=C1
Boiling Point 
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)