Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenyl Chlorodithioformate 97.0+%, TCI America™
SDP

Catalog No. P12665G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P12665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P12665G Supplier TCI America Supplier No. P12665G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 16911-89-0
Molecular Formula C7H5ClS2
Molecular Weight (g/mol) 188.687
MDL Number MFCD00013429
InChI Key XEXIHCWFFNGQDE-UHFFFAOYSA-N
Synonym Chlorodithioformic Acid Phenyl Ester
PubChem CID 592675
IUPAC Name phenyl chloromethanedithioate
SMILES C1=CC=C(C=C1)SC(=S)Cl

Specifications

Boiling Point 95°C
Quantity 5 g
UN Number 3265
Formula Weight 188.69
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Phenyl Chlorodithioformate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.