CAS RN 16919-19-0
Ammonium hexafluorosilicate, 98%
CAS: 16919-19-0 Molecular Formula: F6H8N2Si Molecular Weight (g/mol): 178.153 MDL Number: MFCD00010887 InChI Key: ITHIMUMYFVCXSL-UHFFFAOYSA-P Synonym: ammonium hexafluorosilicate,ammonium fluosilicate,ammonium fluorosilicate,ammonium silicofluoride,cryptohalite,caswell no. 043,diammonium fluosilicate,diammonium fluorosilicate,ammonium silicon fluoride,diammonium hexafluorosilicate PubChem CID: 28145 IUPAC Name: diazanium;hexafluorosilicon(2-) SMILES: [NH4+].[NH4+].F[Si-2](F)(F)(F)(F)F
Ammonium hexafluorosilicate, Puratronic™, 99.999% (metals basis)
CAS: 16919-19-0 Molecular Formula: F6H8N2Si Molecular Weight (g/mol): 178.153 MDL Number: MFCD00010887 InChI Key: ITHIMUMYFVCXSL-UHFFFAOYSA-P Synonym: ammonium hexafluorosilicate,ammonium fluosilicate,ammonium fluorosilicate,ammonium silicofluoride,cryptohalite,caswell no. 043,diammonium fluosilicate,diammonium fluorosilicate,ammonium silicon fluoride,diammonium hexafluorosilicate PubChem CID: 28145 IUPAC Name: diazanium;hexafluorosilicon(2-) SMILES: [NH4+].[NH4+].F[Si-2](F)(F)(F)(F)F
Ammonium hexafluorosilicate, 99.999%, (trace metal basis)
CAS: 16919-19-0 Molecular Formula: H8F6N2Si Molecular Weight (g/mol): 178.15 InChI Key: ITHIMUMYFVCXSL-UHFFFAOYSA-P Synonym: ammonium hexafluorosilicate,ammonium fluosilicate,ammonium fluorosilicate,ammonium silicofluoride,cryptohalite,caswell no. 043,diammonium fluosilicate,diammonium fluorosilicate,ammonium silicon fluoride,diammonium hexafluorosilicate PubChem CID: 28145 IUPAC Name: diazanium;hexafluorosilicon(2-) SMILES: [NH4+].[NH4+].F[Si-2](F)(F)(F)(F)F
VeriSpec™ Silicon Standard for ICP 1000 ppm in H2O, Ricca Chemical
CAS: 16919-19-0 Molecular Formula: F6H8N2Si Molecular Weight (g/mol): 178.153 InChI Key: ITHIMUMYFVCXSL-UHFFFAOYSA-P PubChem CID: 28145 IUPAC Name: diazanium;hexafluorosilicon(2-) SMILES: [NH4+].[NH4+].F[Si-2](F)(F)(F)(F)F
Silicon Standard, 1000mg/L in H2O/0.24 F (Assurance Grade), SPEX CertiPrep™
CAS: 16919-19-0 Molecular Formula: F6H8N2Si Molecular Weight (g/mol): 178.153 InChI Key: ITHIMUMYFVCXSL-UHFFFAOYSA-P PubChem CID: 28145 IUPAC Name: diazanium;hexafluorosilicon(2-) SMILES: [NH4+].[NH4+].F[Si-2](F)(F)(F)(F)F
Silicon ICP Standard, 1000 ppm Si in H2O/tr HF, Ricca Chemical
CAS: 16919-19-0 Molecular Formula: F6H8N2Si Molecular Weight (g/mol): 178.153 InChI Key: ITHIMUMYFVCXSL-UHFFFAOYSA-P PubChem CID: 28145 IUPAC Name: diazanium;hexafluorosilicon(2-) SMILES: [NH4+].[NH4+].F[Si-2](F)(F)(F)(F)F
VeriSpec™ Silicon Standard for ICP 10 g/L in H2O, Ricca Chemical
CAS: 16919-19-0 Molecular Formula: F6H8N2Si Molecular Weight (g/mol): 178.153 InChI Key: ITHIMUMYFVCXSL-UHFFFAOYSA-P PubChem CID: 28145 IUPAC Name: diazanium;hexafluorosilicon(2-) SMILES: [NH4+].[NH4+].F[Si-2](F)(F)(F)(F)F
VeriSpec™ Silicon Standard for ICP/MS 10 ppm in H2O, Ricca Chemical
CAS: 16919-19-0 Molecular Formula: F6H8N2Si Molecular Weight (g/mol): 178.153 InChI Key: ITHIMUMYFVCXSL-UHFFFAOYSA-P PubChem CID: 28145 IUPAC Name: diazanium;hexafluorosilicon(2-) SMILES: [NH4+].[NH4+].F[Si-2](F)(F)(F)(F)F
Silicon ICP Standard, 1000 ppm Si in 3% HNO3, Ricca Chemical
CAS: 16919-19-0 Molecular Formula: F6H8N2Si Molecular Weight (g/mol): 178.153 InChI Key: ITHIMUMYFVCXSL-UHFFFAOYSA-P PubChem CID: 28145 IUPAC Name: diazanium;hexafluorosilicon(2-) SMILES: [NH4+].[NH4+].F[Si-2](F)(F)(F)(F)F
VeriSpec™ Silicon Standard for ICP/MS 100 ppm in H2O, Ricca Chemical
CAS: 16919-19-0 Molecular Formula: F6H8N2Si Molecular Weight (g/mol): 178.153 InChI Key: ITHIMUMYFVCXSL-UHFFFAOYSA-P PubChem CID: 28145 IUPAC Name: diazanium;hexafluorosilicon(2-) SMILES: [NH4+].[NH4+].F[Si-2](F)(F)(F)(F)F
Silicon ICP Standard, 10,000 ppm Si in H2O/tr HF, Ricca Chemical
CAS: 16919-19-0 Molecular Formula: F6H8N2Si Molecular Weight (g/mol): 178.153 InChI Key: ITHIMUMYFVCXSL-UHFFFAOYSA-P PubChem CID: 28145 IUPAC Name: diazanium;hexafluorosilicon(2-) SMILES: [NH4+].[NH4+].F[Si-2](F)(F)(F)(F)F
VeriSpec™ Silicon (Si) Standard for AAS 1000 ppm in H2O, Ricca Chemical
CAS: 16919-19-0 Molecular Formula: F6H8N2Si Molecular Weight (g/mol): 178.153 InChI Key: ITHIMUMYFVCXSL-UHFFFAOYSA-P PubChem CID: 28145 IUPAC Name: diazanium;hexafluorosilicon(2-) SMILES: [NH4+].[NH4+].F[Si-2](F)(F)(F)(F)F
Silicon ICP-MS Standard, 1000 ppm Si in H2O/tr HF, Ricca Chemical
CAS: 16919-19-0 Molecular Formula: F6H8N2Si Molecular Weight (g/mol): 178.153 InChI Key: ITHIMUMYFVCXSL-UHFFFAOYSA-P PubChem CID: 28145 IUPAC Name: diazanium;hexafluorosilicon(2-) SMILES: [NH4+].[NH4+].F[Si-2](F)(F)(F)(F)F
Silicon Standard For ICP, 1000μg/mL in H2O/0.24 F, Assurance Grade, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Ammonium Hexafluorosilicate 98%, Thermo Scientific™
CAS: 16919-19-0 Molecular Formula: F6H8N2Si Molecular Weight (g/mol): 178.153 InChI Key: ITHIMUMYFVCXSL-UHFFFAOYSA-P Synonym: ammonium hexafluorosilicate,ammonium fluosilicate,ammonium fluorosilicate,ammonium silicofluoride,cryptohalite,caswell no. 043,diammonium fluosilicate,diammonium fluorosilicate,ammonium silicon fluoride,diammonium hexafluorosilicate PubChem CID: 28145 IUPAC Name: diazanium;hexafluorosilicon(2-) SMILES: [NH4+].[NH4+].F[Si-2](F)(F)(F)(F)F