Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Hexafluorosilicate 98%, Thermo Scientific™

Catalog No. AC310932500
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC310932500 250 g
AC310930250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC310932500 Supplier Thermo Scientific Chemicals Supplier No. 310932500

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16919-19-0
Molecular Formula F6H8N2Si
Molecular Weight (g/mol) 178.153
InChI Key ITHIMUMYFVCXSL-UHFFFAOYSA-P
Synonym ammonium hexafluorosilicate, ammonium fluosilicate, ammonium fluorosilicate, ammonium silicofluoride, cryptohalite, caswell no. 043, diammonium fluosilicate, diammonium fluorosilicate, ammonium silicon fluoride, diammonium hexafluorosilicate
PubChem CID 28145
IUPAC Name diazanium;hexafluorosilicon(2-)
SMILES [NH4+].[NH4+].F[Si-2](F)(F)(F)(F)F

Specifications

Color White
Physical Form Slightly Adhering Crystalline Powder
Quantity 250 g
Merck Index 15, 524
Formula Weight 178.14
Percent Purity ≥97.5% (ex F)
Density 2.01
Chemical Name or Material Ammonium hexafluorosilicate, 98%
EINECSNumber 240-968-3
RTECSNumber VV7800000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.