accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1692-15-5

IUPAC Name: (pyridin-4-yl)boronic acid
Synonyms: pyridin-4-ylboronic acid
Molecular Weight (g/mol): 122.92
Molecular Formula: C5H6BNO2
InChi Key: QLULGIRFKAWHOJ-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=NC=C1
Molecular Weight (g/mol) 
  • (4)
  • (3)
Quantity 
  • (3)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (4)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)