Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Pyridylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. P15945G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P1594-5G 5 g
P15941G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P1594-5G Supplier TCI America Supplier No. P15945G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1692-15-5
Molecular Formula C5H6BNO2
Molecular Weight (g/mol) 122.92
MDL Number MFCD01074545
InChI Key QLULGIRFKAWHOJ-UHFFFAOYSA-N
PubChem CID 2734379
IUPAC Name (pyridin-4-yl)boronic acid
SMILES OB(O)C1=CC=NC=C1

Specifications

Quantity 5 g
Synonym pyridine-4-boronic acid, 4-pyridineboronic acid, 4-pyridylboronic acid, pyridin-4-yl boronic acid, 4-pyridinylboronic acid
Formula Weight 122.92
Physical Form Crystalline Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.