CAS RN 16941-12-1
IUPAC Name:
dihydrogen hexachloroplatinumdiuide
Synonyms:
chloroplatinic acid
Molecular Weight (g/mol):
415.85
Molecular Formula:
Cl6H8Pt
InChi Key:
QGXPJOUUIMLZTA-UHFFFAOYSA-P
SMILES:
[H+].[H+].Cl.Cl.Cl.Cl.Cl.Cl.[Pt--]
1
–
3
of
3
results
1
Dihydrogen hexachloroplatinate(IV) solution, Pt 20% (cont. Pt)
CAS: 16941-12-1 MDL Number: MFCD00011330
2
Sigma Aldrich Chloroplatinic Acid Solution
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
3
Platinum (IV) Chloride Solution (10% w/v), Fisher Chemical
CAS: 16941-12-1 Molecular Formula: H2Cl6Pt ·xH2O MDL Number: MFCD00011330 Synonym: Platinic Chloride Solution