Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Chloroplatinic Acid Solution

Catalog No. 26258750ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 mL
50 mL
250 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26-258-750ML 50 mL
26-258-710ML 10 mL
26-258-7250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 26-258-750ML Supplier Sigma Aldrich Supplier No. 26258750ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Catalyst precursor for the reaction of silyl hydrides with olefins, hydrosilylation.Also used for the determination of potassium.

Specifications

CAS 16941-12-1
Density 1.05 g/mL at 25°C
Molecular Formula H2Cl6Pt
Linear Formula H2PtCl6
MDL Number MFCD00011330
Quantity 50 mL
Synonym Hexachloroplatinic acid solution; Hydrogen hexachloroplatinate(IV) solution; Platinic chloride solution; Platinum(IV) chloride solution
Molecular Weight (g/mol) 409.81 g/mol
Biological Activity Kidney
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.