accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 169545-27-1

IUPAC Name: 2-(3-{[1,1'-biphenyl]-4-yl}-2-[1-(3,5-dimethylphenyl)-N-methylformamido]propanamido)-3-(1H-indol-3-yl)propanoic acid
Synonyms: 2-(3-{[1,1'-biphenyl]-4-yl}-2-[1-(3,5-dimethylphenyl)-N-methylformamido]propanamido)-3-(1H-indol-3-yl)propanoic acid
Molecular Weight (g/mol): 573.69
Molecular Formula: C36H35N3O4
InChi Key: UZDORQWMYRRLQV-UHFFFAOYNA-N
SMILES: CN(C(CC1=CC=C(C=C1)C1=CC=CC=C1)C(=O)NC(CC1=CNC2=CC=CC=C12)C(O)=O)C(=O)C1=CC(C)=CC(C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results