accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 169590-42-5

IUPAC Name: 4-[5-(4-methylphenyl)-3-(trifluoromethyl)-1H-pyrazol-1-yl]benzene-1-sulfonamide
Synonyms: celecoxib pharmacia spain brand of celecoxib pharmacia brand of celecoxib heumann brand of celecoxib celecoxib product (product) mack brand of celecoxib celebrex celecoxib product celecoxib preparation searle brand of celecoxib
Molecular Weight (g/mol): 381.37
Molecular Formula: C17H14F3N3O2S
InChi Key: RZEKVGVHFLEQIL-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)C1=CC(=NN1C1=CC=C(C=C1)S(N)(=O)=O)C(F)(F)F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results