CAS RN 16960-16-0
CAS RN 16960-16-0
IUPAC Name:
(2S)-1-[(2S)-2-[(2S)-2-[(2S)-6-amino-2-[(2S)-2-{[(2S)-1-[(2R)-2-[(2R)-2-[(2R)-6-amino-2-[(2R)-6-amino-2-{2-[(2S)-2-{[(2S)-1-[(2S)-6-amino-2-{2-[(2S)-2-[(2S)-2-[(2S)-2-[(2S)-2-[(2S)-2-[(2S)-2-[(2S)-2-[(2S)-2-(2-amino-3-hydroxypropanamido)-3-(4-hydroxyphenyl)propanamido]-3-hydroxypropanamido]-4-(methylsulfanyl)butanamido]-4-carboxybutanamido]-3-(1H-imidazol-4-yl)propanamido]-3-phenylpropanamido]-5-carbamimidamidopentanamido]-3-(1H-indol-3-yl)propanamido]acetamido}hexanoyl]pyrrolidin-2-yl]formamido}-3-methylbutanamido]acetamido}hexanamido]hexanamido]-5-carbamimidamidopentanamido]-5-carbamimidamidopentanoyl]pyrrolidin-2-yl]formamido}-3-methylbutanamido]hexanamido]-3-methylbutanamido]-3-(4-hydroxyphenyl)propanoyl]pyrrolidine-2-carboxylic acid
Synonyms:
synacthen synthetic acth tetracosactrin acthα1-24 tetracosapeptide α 1-24 corticotropin 1-24-corticotropin α1-24-corticotropin tetracosactid acth, synthetic
Molecular Weight (g/mol):
2933.49
Molecular Formula:
C136H210N40O31S
InChi Key:
ZOEFCCMDUURGSE-CQVUSSRSSA-N
SMILES:
CSCC[C@H](NC(=O)[C@H](CO)NC(=O)[C@H](CC1=CC=C(O)C=C1)NC(=O)C(N)CO)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](CC1=CNC=N1)C(=O)N[C@@H](CC1=CC=CC=C1)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CC1=CNC2=CC=CC=C12)C(=O)NCC(=O)N[C@@H](CCCCN)C(=O)N1CCC[C@H]1C(=O)N[C@@H](C(C)C)C(=O)NCC(=O)N[C@H](CCCCN)C(=O)N[C@H](CCCCN)C(=O)N[C@H](CCCNC(N)=N)C(=O)N[C@H](CCCNC(N)=N)C(=O)N1CCC[C@H]1C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](CC1=CC=C(O)C=C1)C(=O)N1CCC[C@H]1C(O)=O
1
–
1
of
1
results
1