Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Adrenocorticotropic Hormone (1-24), human

Catalog No. AAJ66365MCR
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66365MCR 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ66365MCR Supplier Thermo Scientific Chemicals Supplier No. J66365MCR
Add to Cart

missing translation for 'validMainframeMsg'

Used in the treatment of West syndrome. It activates G proteins. It stimulates secretion of glucocorticoid steroid hormones from adrenal cortex cells. It also enhances transcription of mitochondrial genes.

Adrenocorticotropic Hormone (1-24), human is used in the treatment of West syndrome. It activates G proteins. It stimulates secretion of glucocorticoid steroid hormones from adrenal cortex cells. It also enhances transcription of mitochondrial genes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16960-16-0

Specifications

Content And Storage Store at -20°C
Format Solid
Quantity 1 mg

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.