accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH4NH4ClClClClClClSn

CAS RN 16960-53-5

IUPAC Name: tin(4+) diammonium hexachloride
Synonyms: tin(4+) diammonium hexachloride
Molecular Weight (g/mol): 367.49
Molecular Formula: Cl6H8N2Sn
InChi Key: UDMAVAHFQCVSSE-UHFFFAOYSA-J
SMILES: [NH4+].[NH4+].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Sn+4]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results