Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium hexachlorostannate(IV), 98%

Catalog No. AAA1515230
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1515230 250 g
AAA1515218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1515230 Supplier Thermo Scientific Chemicals Supplier No. A1515230
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 16960-53-5 | Cl6H8N2Sn | 367.488 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16960-53-5
Molecular Formula Cl6H8N2Sn
Molecular Weight (g/mol) 367.488
MDL Number MFCD00036340
InChI Key UDMAVAHFQCVSSE-UHFFFAOYSA-J
Synonym ammonium tin chloride, stannic ammonium chloride, ammonium hexachlorostannate, diammonium hexachlorostannate, ammonium hexachlorostannate 2-, diammonium hexachlorostannate 2-, ammonium tin 4+ chloride 2:1:6, diazanium tin 4+ hexachloride, stannate 2-, hexachloro-, diammonium, oc-6-11, stannate 2-, hexachloro-, ammonium 1:2 , oc-6-11
PubChem CID 167595
IUPAC Name diazanium;tin(4+);hexachloride
SMILES [NH4+].[NH4+].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Sn+4]

Specifications

Melting Point (decomposition)
Quantity 250 g
Formula Weight 367.48
Percent Purity 98%
Density 2.4
Chemical Name or Material Ammonium hexachlorostannate(IV)
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 241-033-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.