accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 169685-29-4

IUPAC Name: 4-(trifluoromethoxy)benzene-1-thiol
Synonyms: 4-(trifluoromethoxy)benzenethiol
Molecular Weight (g/mol): 194.17
Molecular Formula: C7H5F3OS
InChi Key: JHVNSRJPBXPZJU-UHFFFAOYSA-N
SMILES: FC(F)(F)OC1=CC=C(S)C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)