Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Trifluoromethoxy)thiophenol, 95%

Catalog No. AAL1867406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1867406 5 g
AAL1867403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1867406 Supplier Thermo Scientific Chemicals Supplier No. L1867406
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 169685-29-4 | C7H5F3OS | 194.171 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 169685-29-4
Molecular Formula C7H5F3OS
Molecular Weight (g/mol) 194.171
MDL Number MFCD00052312
InChI Key JHVNSRJPBXPZJU-UHFFFAOYSA-N
Synonym 4-trifluoromethoxy thiophenol, 4-trifluoromethoxy benzenethiol, 4-trifluoromethoxy benzene-1-thiol, 4-trifluoromethoxythiophenol, benzenethiol, 4-trifluoromethoxy, pubchem15268, acmc-1bxy9, 4-trifluoromethoxy-thiophenol, p-trifluoromethoxybenzenethiol
PubChem CID 2777360
IUPAC Name 4-(trifluoromethoxy)benzenethiol
SMILES C1=CC(=CC=C1OC(F)(F)F)S

Specifications

Boiling Point 64°C to 66°C (15 mmHg)
Refractive Index 1.475
Quantity 5 g
UN Number UN2810
Beilstein 7916918
Sensitivity Air Sensitive
Formula Weight 194.18
Percent Purity 95%
Chemical Name or Material 4-(Trifluoromethoxy)thiophenol
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.