accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 169870-82-0

CH3CH3H3COHONHOOCH3CH3H3C

CAS RN 169870-82-0

IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}-3,3-dimethylbutanoic acid
Synonyms: 2-[(tert-butoxycarbonyl)amino]-3,3-dimethylbutanoic acid
Molecular Weight (g/mol): 231.29
Molecular Formula: C11H21NO4
InChi Key: LRFZIPCTFBPFLX-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(C(O)=O)C(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results