accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONH3COCH3OCH3OH3C

CAS RN 1699-51-0

IUPAC Name: 1-[(3,4-dimethoxyphenyl)methyl]-6,7-dimethoxy-2-methyl-1,2,3,4-tetrahydroisoquinoline
Synonyms: laudanosine
Molecular Weight (g/mol): 357.45
Molecular Formula: C21H27NO4
InChi Key: KGPAYJZAMGEDIQ-UHFFFAOYNA-N
SMILES: COC1=CC=C(CC2N(C)CCC3=CC(OC)=C(OC)C=C23)C=C1OC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results