Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Laudanosine 99%, Thermo Scientific™

Catalog No. AC147360010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC147360010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC147360010 Supplier Thermo Scientific Chemicals Supplier No. 147360010

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1699-51-0
Molecular Formula C21H27NO4
Molecular Weight (g/mol) 357.45
MDL Number MFCD00006910
InChI Key KGPAYJZAMGEDIQ-UHFFFAOYNA-N
Synonym laudanosine, dl-laudanosine, +--laudanosine, r,s-laudanosine, laudanosine r,s, 1-3,4-dimethoxyphenyl methyl-6,7-dimethoxy-2-methyl-1,2,3,4-tetrahydroisoquinoline, +--1,2,3,4-tetrahydro-6,7-dimethoxy-2-methyl-1-veratrylisoquinoline, isoquinoline, 1,2,3,4-tetrahydro-6,7-dimethoxy-2-methyl-1-veratryl-, +-, 1-1-3,4-dimethoxyphenyl methyl-1,2,3,4-tetrahydro-6,7-dimethoxy-2-methylisoquinoline, 1-3,4-dimethoxyphenyl methyl-6,7-dimethoxy-2-methyl-1,2,3,4-tetrahydroisoqu inoline
PubChem CID 15548
IUPAC Name 1-[(3,4-dimethoxyphenyl)methyl]-6,7-dimethoxy-2-methyl-1,2,3,4-tetrahydroisoquinoline
SMILES COC1=CC=C(CC2N(C)CCC3=CC(OC)=C(OC)C=C23)C=C1OC

Specifications

Melting Point 113°C to 117°C
Color Yellow
Infrared Spectrum Authentic
Quantity 1 g
Merck Index 10, 5211
Formula Weight 357.45
Percent Purity ≥98.5%
Physical Form Fine Crystalline Powder
Chemical Name or Material DL-Laudanosine, 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.