accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16993-86-5

IUPAC Name: 2-methyloct-2-ene
Synonyms: 2-methyloct-2-ene
Molecular Weight (g/mol): 126.24
Molecular Formula: C9H18
InChi Key: YBOZNTGUYASNRA-UHFFFAOYSA-N
SMILES: CCCCCC=C(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results