Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-2-octene, MP Biomedicals

Catalog No. ICN225252 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN225252 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN225252 Supplier MP Biomedicals, Inc Supplier No. 0522525210

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 16993-86-5
Molecular Formula C9H18
Molecular Weight (g/mol) 126.243
InChI Key YBOZNTGUYASNRA-UHFFFAOYSA-N
Synonym 2-methyl-2-octene, 2-octene, 2-methyl
PubChem CID 140163
IUPAC Name 2-methyloct-2-ene
SMILES CCCCCC=C(C)C

Specifications

Quantity 10 g
Formula Weight 126.24
Chemical Name or Material 2-Methyl-2-Octene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.