accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 170142-20-8

IUPAC Name: 7-chloro-2-(4-methoxy-2-methylphenyl)-1H,2H,3H,4H,5H,10H-pyridazino[4,5-b]quinoline-1,4,10-trione
Synonyms: 7-chloro-2-(4-methoxy-2-methylphenyl)-3H,5H-pyridazino[4,5-b]quinoline-1,4,10-trione
Molecular Weight (g/mol): 383.79
Molecular Formula: C19H14ClN3O4
InChi Key: PSTDMIAVUUYOOQ-UHFFFAOYSA-N
SMILES: COC1=CC(C)=C(C=C1)N1NC(=O)C2=C(C1=O)C(=O)C1=CC=C(Cl)C=C1N2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results