Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZD 9379, Tocris Bioscience™
SDP

Catalog No. 332210 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-221-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 33-221-0 Supplier R&D Systems Supplier No. 3322/10

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Brain penetrant NMDA antagonist; acts at glycine site

NMDA receptor antagonist; acts at glycine site. Neuroprotective; reduces infarct size and frequency of spreading depressions in a rat model of ischemic stroke. Brain penetrant.

Chemical Identifiers

CAS 170142-20-8
Molecular Formula C19H14ClN3O4
Molecular Weight (g/mol) 383.79
InChI Key PSTDMIAVUUYOOQ-UHFFFAOYSA-N
Synonym pstdmiavuuyooq-uhfffaoysa-n, 7-chloro-4-hydroxy-2-4-methoxy-2-methylphenyl-1,2,5,10-tetrahydropyridazino 4,5-b quinoline-1,10-dione, 7-chloro-2,3-dihydro-2-4-methoxy-2-methylphenyl pyridazino 4,5-b quinoline-1,4,10 5h trione, 7-chloro-4-hydroxy-2-4-methoxy-2-methylphenyl-1,2,5,10-tetrahydropyridazino-4,5-b quinoline-1,10-dione
PubChem CID 9843028

Specifications

Quantity 10 mg
Formula Weight 383.79
Chemical Name or Material ZD 9379
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.