accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OCH3HS

CAS RN 17042-24-9

IUPAC Name: 2-sulfanylpentan-3-one
Synonyms: 2-sulfanylpentan-3-one
Molecular Weight (g/mol): 118.19
Molecular Formula: C5H10OS
InChi Key: TWOGJUHCCNLYPC-UHFFFAOYNA-N
SMILES: CCC(=O)C(C)S

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results