Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Mercapto-3-pentanone 95.0+%, TCI America™
SDP

Catalog No. M25155G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M25155G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M25155G Supplier TCI America Supplier No. M25155G

Chemical Identifiers

CAS 17042-24-9
Molecular Formula C5H10OS
Molecular Weight (g/mol) 118.194
InChI Key TWOGJUHCCNLYPC-UHFFFAOYSA-N
PubChem CID 529635
IUPAC Name 2-sulfanylpentan-3-one
SMILES CCC(=O)C(C)S

Specifications

Color Yellow
Boiling Point 49°C
Quantity 5 g
UN Number 3336
Formula Weight 118.19
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Mercapto-3-pentanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.