accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1704465-79-1

IUPAC Name: N3,N3,N6,N6-tetramethylacridine-3,6-diamine hydrochloride
Synonyms: basic orange 14
Molecular Weight (g/mol): 301.82
Molecular Formula: C17H20ClN3
InChi Key: VSTHNGLPHBTRMB-UHFFFAOYSA-N
SMILES: Cl.CN(C)C1=CC2=NC3=CC(=CC=C3C=C2C=C1)N(C)C
Color 
  • (1)
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)