Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Acridine Orange, hydrochloride hydrate, pure, ca. 80 % dye content

Catalog No. AC423340010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC423340010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC423340010 Supplier Thermo Scientific Chemicals Supplier No. 423340010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

CAS: 1704465-79-1 | C17H20ClN3 | 301.82 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1704465-79-1
Molecular Formula C17H20ClN3
Molecular Weight (g/mol) 301.82
MDL Number MFCD00150070
InChI Key VSTHNGLPHBTRMB-UHFFFAOYSA-N
Synonym Basic orange 14, 3, 6-Bis(dimethylamino)acridine hydrochloride, C.I. 46005
PubChem CID 517204
ChEBI CHEBI:51739
IUPAC Name N3,N3,N6,N6-tetramethylacridine-3,6-diamine hydrochloride
SMILES Cl.CN(C)C1=CC2=NC3=CC(=CC=C3C=C2C=C1)N(C)C

Specifications

Infrared Spectrum Authentic
Loss on Drying 5% max. (105°C)
Packaging Glass bottle
UV_Vis 75% min.
Beilstein 22, 487
Color Brown to Orange
Quantity 1 g
Physical Form Powder
Formula Weight 301.82
Grade Extra Pure
Chemical Name or Material Acridine orange, hydrochloride hydrate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS P Statement
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Suspected of causing genetic defects.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF ON SKIN: Wash with plenty of soap and water.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.