accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COONCO

CAS RN 17046-22-9

IUPAC Name: butyl 2-isocyanatoacetate
Synonyms: butyl 2-isocyanatoacetate
Molecular Weight (g/mol): 157.17
Molecular Formula: C7H11NO3
InChi Key: RMZSOGJUEUFCBK-UHFFFAOYSA-N
SMILES: CCCCOC(=O)CN=C=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results