Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butyl Isocyanatoacetate 97.0+%, TCI America™
SDP

Catalog No. I03585G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I03585G 5 g
I035825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I03585G Supplier TCI America Supplier No. I03585G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 17046-22-9
Molecular Formula C7H11NO3
Molecular Weight (g/mol) 157.17
MDL Number MFCD00041743
InChI Key RMZSOGJUEUFCBK-UHFFFAOYSA-N
Synonym butyl isocyanatoacetate, n-butyl isocyanatoacetate, butyl n-oxomethylene glycinate, acetic acid, isocyanato-, butyl ester, acetic acid, 2-isocyanato-, butyl ester, unii-cvm3w9q955, isocyanatoacetic acid n-butyl ester, butyl=isocyanatoacetate, butyl n-oxomethylidene glycinate
PubChem CID 86921
IUPAC Name butyl 2-isocyanatoacetate
SMILES CCCCOC(=O)CN=C=O

Specifications

Color Yellow
Boiling Point 79°C
Quantity 5 g
UN Number 2206
Formula Weight 157.17
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Butyl Isocyanatoacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.