accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1705581-28-7

IUPAC Name: 5-aminopentanoic acid hydroiodide
Synonyms: 5-aminovaleric acid hydroiodide
Molecular Weight (g/mol): 245.06
Molecular Formula: C5H12INO2
InChi Key: QRCPJIVRDACIKP-UHFFFAOYSA-N
SMILES: I.NCCCCC(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results