Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Aminovaleric Acid Hydroiodide (Low water content) 97.0+%, TCI America™
SDP

Catalog No. A29845G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A29845G 5 g
A29841G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A29845G Supplier TCI America Supplier No. A29845G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1705581-28-7
Molecular Formula C5H12INO2
Molecular Weight (g/mol) 245.06
InChI Key QRCPJIVRDACIKP-UHFFFAOYSA-N
Synonym 5-Aminopentanoic Acid Hydroiodide, Homopiperidinic Acid Hydroiodide, 5-AVAI
PubChem CID 118324268
IUPAC Name 5-aminopentanoic acid;hydroiodide
SMILES C(CCN)CC(=O)O.I

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 245.06
Percent Purity ≥97.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Aminovaleric Acid Hydroiodide (Low water content)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.