accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClCl

CAS RN 17060-07-0

IUPAC Name: 1,2-dichloroethane
Synonyms: ethylene dichloride 1,2-bichloroethane caswell no. 440 di-chlor-mulsion RY dichloro-1,2-ethane DCE EDC bichlorure d'ethylene dutch oil epa pesticide chemical code 042003
Molecular Weight (g/mol): 98.95
Molecular Formula: C2H4Cl2
InChi Key: WSLDOOZREJYCGB-UHFFFAOYSA-N
SMILES: ClCCCl
Quantity 
  • (1)
  • (1)
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results