accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClH3CONClNH2NH2Cl

CAS RN 17070-45-0

IUPAC Name: 6-chloro-N-{3-[(2-chloroethyl)azaniumyl]propyl}-2-methoxyacridin-9-aminium dichloride
Synonyms: 6-chloro-N-{3-[(2-chloroethyl)ammonio]propyl}-2-methoxyacridin-9-aminium dichloride
Molecular Weight (g/mol): 451.21
Molecular Formula: C19H23Cl4N3O
InChi Key: LMEMIKWTNPWYMI-UHFFFAOYSA-N
SMILES: [Cl-].[Cl-].COC1=CC=C2N=C3C=C(Cl)C=CC3=C([NH2+]CCC[NH2+]CCCl)C2=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results