Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-chloro-9-(3-N-(2-chloroethyl-amino)propylamino-2-methoxyacridine dihydrochloride, Thermo Scientific™

Catalog No. AC252211000
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC252211000 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC252211000 Supplier Thermo Scientific Chemicals Supplier No. 252211000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17070-45-0
Molecular Formula C19H23Cl4N3O
Molecular Weight (g/mol) 451.213
MDL Number MFCD00013320
InChI Key LMEMIKWTNPWYMI-UHFFFAOYSA-N
Synonym acridine mutagen icr 191, icr-191 dihydrochloride, 6-chloro-9-3-2-chloroethylamino propylamino-2-methoxyacridine dihydrochloride, unii-9u6a4q2be4, n-2-chloroethyl-n'-6-chloro-2-methoxyacridin-9-yl propane-1,3-diamine dihydrochloride, n1-6-chloro-2-methoxyacridin-9-yl-n3-2-chloroethyl propane-1,3-diamine dihydrochloride, 2-methoxy-6-chloro-9-2-chloroethylaminopropylamino acridine dihydrochloride, 6-chloro-n-3-2-chloroethyl amino propyl-2-methoxyacridin-9-amine dihydrochloride, ames mutagen 191, icr 191 dihydrochloride
PubChem CID 104809
IUPAC Name N-(2-chloroethyl)-N'-(6-chloro-2-methoxyacridin-9-yl)propane-1,3-diamine;dihydrochloride
SMILES COC1=CC2=C(C3=C(C=C(C=C3)Cl)N=C2C=C1)NCCCNCCCl.Cl.Cl

Specifications

Color Yellow
Infrared Spectrum Authentic
Quantity 100 mg
Formula Weight 451.24
Physical Form Crystals or Powder
Chemical Name or Material 6-chloro-9-(3-N-(2-chloroethyl-amino)propylamino-2-methoxyacridine dihydrochloride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Suspected of causing cancer.
Toxic in contact with skin.
Fatal if inhaled.
Toxic if swallowed.
Suspected of causing genetic defects.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.