CAS RN 170713-75-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Nociceptin
Endogenous ligand for the opioid-like G-protein coupled receptor, ORL1 (Opioid Receptor-Like 1).